Learn More
Hughes Jacketed and Insulated Tank Shower
Product Code. p-7227320
Change view
Click to view available options
Capacity (Metric):
350 L
1500 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
17772348 350 L Each
17762348 1500 L Each
2 options
Product Code. 17772348
Brand: Hughes HSHJ14K3SS
Only null left
Add to basket
Add to basket
Insulated emergency tank shower with eye/face wash unit, for use in non-hazardous hot climates. Designed to meet and exceed ANSI standards by providing tepid water, even when there is no mains water supply.
  • Insulated emergency tank shower with eye/face wash unit, for use in non-hazardous hot climates
  • Jacketed and insulated models maintain a steady temperature
  • Suitable for use where insufficient water pressure exists
  • Side mounted eye/face wash unit
  • Designed to meet and exceed ANSI standards by providing tepid water for over 15 minutes at a guaranteed flow of 76 liters per minute, even when there is no mains water supply
Specifications
Capacity (Metric) 350 L
Material Stainless Steel
Product Type Tank Shower