Learn More
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and Plain Arm
Product Code. 15844734
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
15844734 250 mL Each
15834734 25 mL Each
15854734 50 mL Each
15814734 100 mL Each
15864734 500 mL Each
15824734 1000 mL Each
6 options
Product Code. 15844734
Brand: GPE Scientific™ CSB/250
Only null left
Add to basket
Add to basket
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and Plain Arm is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
  • Designed with a side arm for evacuating or filling with gases
  • For connection to Schlenk lines
  • Round-bottomed shape
  • Includes plain arm
  • Includes O-ring tap
Specifications
Capacity (Metric) 250 mL
Includes O-Ring Tap and Plain Arm
Product Type Vacuum/Schlenk Flask