Learn More
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup
Product Code. 15824744
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
15824744 500 mL Each
15894734 25 mL Each
15814744 50 mL Each
15874734 100 mL Each
15804744 250 mL Each
15884734 1000 mL Each
6 options
Product Code. 15824744
Brand: GPE Scientific™ CSR/500
Only null left
Add to basket
Add to basket
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
  • Designed with a side arm for evacuating or filling with gases
  • For connection to Schlenk lines
  • Round-bottomed shape
  • Includes O-ring tap
  • Includes O-ring ball joint cup
Specifications
Capacity (Metric) 500 mL
Includes O-Ring Tap and O-Ring Ball Joint Cup
Product Type Vacuum/Schlenk Flask
Side Arm Joint Size: 29