Learn More
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Glass Key and Socket
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Description
- Includes glass key and socket
- Available in various capacities
- Available with different joint sizes
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Includes | Glass key and socket |
| Product Type | Schlenk Flask |
| Socket Joint | Joint size: 24 |
| Bottom Shape | Round |