Learn More
GPE Scientific™ J Young Schlenk Flask, Cylindrical with Fine Thread Tap and Cone
Product Code. 15854784
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
15854784 100 mL Each
15768029 25 mL Each
15778029 50 mL Each
15884784 250 mL Each
15894784 500 mL Each
15874784 1000 mL Each
6 options
Product Code. 15854784
Brand: GPE Scientific™ JCC/100
Only null left
Add to basket
Add to basket
GPE Scientific™ J Young Schlenk Flask, Cylindrical with Fine Thread Tap and Cone is specially designed for use with vacuum manifolds and Schlenk lines.
  • Solidly constructed for use with vacuum manifolds
  • Designed for use with Schlenk lines
  • Includes fine thread tap and cone
Specifications
Capacity (Metric) 100 mL
For Use With (Equipment) Vacuum manifolds and Schlenk lines
Includes Fine thread tap and cone
Product Type Schlenk Flask
Socket Joint Size: 24