Learn More
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
1325.00 NOK - 1660.00 NOK
Specifications
Includes | O-ring tap and cold finger plain arm |
---|---|
Product Type | Sample Flask |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15854744
|
GPE Scientific™
CSY/25 |
25 mL |
1325.00 NOK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15874744
|
GPE Scientific™
CSY/50 |
50 mL |
1390.00 NOK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15834744
|
GPE Scientific™
CSY/100 |
100 mL |
1430.00 NOK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15864744
|
GPE Scientific™
CSY/250 |
250 mL |
1445.00 NOK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15884744
|
GPE Scientific™
CSY/500 |
500 mL |
1530.00 NOK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15844744
|
GPE Scientific™
CSY/1000 |
1000 mL |
1660.00 NOK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
Description
- Designed with a side arm for evacuating or filling with gases
- For connection to Schlenk lines
- Includes O-ring tap
- Includes cold finger plain arm
Specifications
O-ring tap and cold finger plain arm | |
Sample Flask |