Learn More
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm
Description
- Designed with a side arm for evacuating or filling with gases
- For connection to Schlenk lines
- Includes O-ring tap
- Includes cold finger plain arm
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Includes | O-ring tap and cold finger plain arm |
| Product Type | Sample Flask |