Learn More
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap and B14 Socket
Product Code. 15814804
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15814804 500 mL Each
15884794 25 mL Each
15804804 50 mL Each
15864794 100 mL Each
15894794 250 mL Each
15874794 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15814804 Supplier GPE Scientific™ Supplier No. JCS/500
Estimated Shipment: 03-02-2027
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap and B14 Socket is specially designed for use with vacuum manifolds and Schlenk lines.
  • Solidly constructed for use with vacuum manifolds
  • Designed for use with Schlenk lines
  • Includes fine thread tap and B14 socket
Specifikationer
Capacity (Metric) 500 mL
For Use With (Equipment) Vacuum manifolds and Schlenk lines
Includes Fine thread tap and B14 socket
Product Type Sample Flask
Socket Joint Size: 14
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.