Learn More
GPE Scientific™ J Young NMR Valve with Tube Attached
Product Code. 15864874
Click to view available options
Type:
NMR Valve with Tube, 200 MHz
NMR Valve with Tube, 500 MHz
Length (English):
7 in.
8 in.
Unit Size:
Each
Product Code. 15864874
Brand: GPE Scientific™ S5200JY8YELLOW
GPE Scientific™ J Young NMR Valve with Tube Attached allows you to handle your NMR sample without flame sealing your tubes.
  • Handle your NMR sample without flame sealing your tubes.
  • Teflon covered O-ring eliminates material incompatibilities.
  • Completely greaseless Teflon assembly means the tube is easy to use and to disassemble for cleaning.
  • Includes female joint for quick attachment to your vacuum rack.
  • Yellow piston
  • Negative pressure/high vacuum.
  • Leak tested to 10-9mbar liters/sec.
  • Internal positive pressure 5bar, 72psi
Specifications
Type NMR Valve with Tube, 200 MHz
Includes 5mm NMR Valve
Length (English) 8 in.