Learn More
GPE Scientific™ J Young Double Oblique Taps, Fast Thread
GPE Scientific™ J Young Double Oblique Taps, Fast Thread are made in various stopcock sizes and with different side arm diameters.
1495.00 NOK
Specifications
Type | J Young double oblique taps with fast threads |
---|---|
Includes | Dual taps |
Product Code | Brand | Plug Bore | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Plug Bore | Price | Quantity & Availability | |||||
15874864
|
GPE Scientific™
PTT/D/6 |
6 mm |
1495.00 NOK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15884864
|
GPE Scientific™
PTT/D/8 |
8 mm |
1495.00 NOK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15864864
|
GPE Scientific™
PTT/D/10 |
10 mm |
1495.00 NOK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
Description
- Fast thread type
- Available in various stopcock sizes
- Available with different side arm diameters
Specifications
J Young double oblique taps with fast threads | |
Dual taps |