Learn More
IYD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16667-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes an enzyme that catalyzes the oxidative NADPH-dependent deiodination of mono- and diiodotyrosine, which are the halogenated byproducts of thyroid hormone production. The N-terminus of the protein functions as a membrane anchor. Mutations in this gene cause congenital hypothyroidism due to dyshormonogenesis type 4, which is also referred to as deiodinase deficiency, or iodotyrosine dehalogenase deficiency, or thyroid hormonogenesis type 4. Alternative splicing results in multiple transcript variants.Specifications
| IYD | |
| Polyclonal | |
| Unconjugated | |
| IYD | |
| C6orf71, DEHAL1, Iodotyrosine dehalogenase 1, iodotyrosine deiodinase, IYD, IYD 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 389434 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PHW0 | |
| IYD | |
| IYD Fusion Protein Ag10146 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |