missing translation for 'onlineSavingsMsg'
Learn More

Ixazomib citrate, MedChemExpress

Product Code. 16426697
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16426697 5 mg 5mg
16493978 10 mg 10mg
16440337 50 mg 50mg
16474986 100 mg 100mg
4 options
This item is not returnable. View return policy

Product Code. 16426697

Brand: Medchem Express HY10452

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ixazomib citrate (MLN9708) is a reversible inhibitor of the chymotrypsin-like proteolytic β5 site of the 20S proteasome with an IC50 of 3.4 nM and a Ki of 0.93 nM.

Ixazomib citrate (MLN9708) is a reversible inhibitor of the chymotrypsin-like proteolytic β5 site of the 20S proteasome with an IC50 of 3.4 nM and a Ki of 0.93 nM.

Specifications

Chemical Name or Material Ixazomib citrate
Physical Form Solid
CAS 1239908-20-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H23BCl2N2O9
For Use With (Application) Cancer-Kinase/protease
Synonym MLN9708
Solubility Information DMSO : 250 mg/mL (483.45 mM; Need ultrasonic)
SMILES O=C(O)CC1(CC(O)=O)OB([C@@H](NC(CNC(C2=CC(Cl)=CC=C2Cl)=O)=O)CC(C)C)OC1=O
Molecular Weight (g/mol) 517.12
Formula Weight 517.12
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.