missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ITGA5/ITGB1 Monoclonal Antibody (05)
GREENER_CHOICE

Product Code. 17136693 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17136693 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17136693 Supplier Invitrogen™ Supplier No. MA536058

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human ITGA5 & ITGB1 Heterodimer.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA5/ITGB1
Applications Flow Cytometry
Classification Monoclonal
Clone 05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ITGA5
Gene Accession No. P05556, P08648
Gene Alias 4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CD49 antigen-like family member E; Cd49e; ENSMUSG00000051907; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; Fibronectin receptor subunit beta; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; FNRB; glycoprotein IIa; Gm9863; gpIIa; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin beta 1; integrin beta 1 (fibronectin receptor beta); Integrin beta1; integrin beta-1; integrin subunit alpha 5; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); Itga5; Itga5_retired; ITGB1; MDF2; MSK12; very late activation protein 5, alpha subunit; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLA5; VLA-5; VLA5A; VLAB; VLA-BETA
Gene Symbols ITGA5, ITGB1
Host Species Mouse
Immunogen Recombinant Human ITGA5 & ITGB1 Heterodimer Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3678, 3688
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.