Learn More
KGW Isotherm™ Isotherm Borosilicate Glass Insulating Lid
Click to view available options
Capacity (Metric):
1000 mL
1500 mL
2000 mL
2500 mL
3000 mL
Unit Size:
Each
Specifications
Specifications
| Capacity (Metric) | 3000 mL |
| Material | Borosilicate Glass |
| For Use With (Application) | Flexible Applications In Laboratory |
| For Use With (Equipment) | Dewar Container; Type 20 Plugs |
| No. per Pack | 1 |
| Product Type | Insulating Lid |