Learn More
Isorhamnetin, MedChemExpress
Product Code. 16421508
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16421508
Brand: Medchem Express HYN0776
MedChemExpress Isorhamnetin is a flavonoid compound extracted from the Chinese herb Hippophae rhamnoides L.. Isorhamnetin suppresses skin cancer through direct inhibition of MEK1 and PI3K.
Isorhamnetin is a flavonoid compound extracted from the Chinese herb Hippophae rhamnoides L.. Isorhamnetin suppresses skin cancer through direct inhibition of MEK1 and PI3K.
Specifications
Chemical Name or Material Isorhamnetin
CAS 480-19-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H12O7
Quantity 5 mg
Synonym 3'-Methylquercetin
Solubility Information DMSO : 100 mg/mL (316.20 mM; Need ultrasonic)
SMILES O=C1C(O)=C(C2=CC=C(O)C(OC)=C2)OC3=CC(O)=CC(O)=C13
Molecular Weight (g/mol) 316.26
Formula Weight 316.26
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.