Learn More
Isoquinoline-3-carboxylic acid hydrate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 203626-75-9 |
| Molecular Formula | C10H9NO3 |
| MDL Number | MFCD00167216 |
| Quantity | 1 g |
| SMILES | O=C(C1=CC2=C(C=N1)C=CC=C2)O.[H]O[H] |
| Molecular Weight (g/mol) | 191.18 |
| Percent Purity | 95% |
| Chemical Name or Material | Isoquinoline-3-carboxylic acid hydrate |