Learn More
Isopropyl Propyl Ether, TRC
Product Code. 30137396
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137396 1 g 1g
30135498 500 mg 500mg
17973035 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137396 Supplier Toronto Research Chemicals Supplier No. TRCI7485001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 627-08-7
Molecular Formula C6 H14 O
Molecular Weight (g/mol) 102.175
Synonym 1-(1-Methylethoxy)propane, 1-(Propan-2-yloxy)propane, Isopropyl propyl ether, Propyl iso-propyl ether, Propyl isopropyl ether;
IUPAC Name 1-propan-2-yloxypropane
SMILES CCCOC(C)C
Spezifikation
Quantity 1 g
InChI Formula InChI=1S/C6H14O/c1-4-5-7-6(2)3/h6H,4-5H2,1-3H3
Formula Weight 102.104
Chemical Name or Material Isopropyl Propyl Ether