Learn More
Phagogene™ Ethanol Spray
Product Code. 11726954
Change view
Click to view available options
Volume (Metric):
750 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11726954 750 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11726954 Supplier Phagogene™ Supplier No. 60416
  • ONLINE EXCLUSIVE PRICE
    656.00 NOK
    537.38 NOK / Pack of 6
    Save 118.62 NOK
    18% Off
 Restricted Item
Estimated Shipment: 11-12-2026
PHAGO'SPRAY DM is a professional disinfectant cleaner for the disinfection of surfaces in high-risk environments. PHAGO'SPRAY DM can be used as a spray on walls, ceilings and furniture in care and surgery departments.
  • Ready to use
  • Broad spectrum disinfectant Efficacy on Rotavirus in 5 minutes
  • A nozzle with two positions (spray and stop pictograms)
  • A removable ergo for use in foam or spray
Specifications
Product Type Disinfectant Cleaner
Form Clear Liquid
Scent Fresh Scent
Sterility Non-sterile
Quantity 6/Pk.
Size 750 mL
Volume (Metric) 750 mL
compliance-icons compliance-icons
Product Identifier
  • PHAGOSPRAY DM
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.