Learn More
Isopilocarpic Acid Sodium Salt, TRC
Product Code. 30132415
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132415 25 mg 25mg
30132764 5 mg 5mg
30041353 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132415 Supplier Toronto Research Chemicals Supplier No. TRCI82196025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101769-87-3
Molecular Formula C11 H17 N2 O3 . Na
Molecular Weight (g/mol) 248.25
Synonym 1H-Imidazole-5-butanoic acid, α-ethyl-β-(hydroxymethyl)-1-methyl-, monosodium salt, [R-(R*,R*)]- (9CI), (2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt, Pilocarpine Hydrochloride Imp. C (EP) as Sodium Salt, Isopilocarpic Acid Sodium Salt
IUPAC Name sodium;(2R,3R)-2-ethyl-3-(hydroxymethyl)-4-(3-methylimidazol-4-yl)butanoate
SMILES [Na+].CC[C@H]([C@H](CO)Cc1cncn1C)C(=O)[O-]
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C11H18N2O3.Na/c1-3-10(11(15)16)8(6-14)4-9-5-12-7-13(9)2;/h5,7-8,10,14H,3-4,6H2,1-2H3,(H,15,16);/q;+1/p-1/t8-,10+;/m0./s1
Formula Weight 248.1137
Percent Purity >95%
Chemical Name or Material (2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Isopilocarpic Acid Sodium Salt)
Grade HPLC