missing translation for 'onlineSavingsMsg'
Learn More

Isoorientin, MedChemExpress

Product Code. p-200098188
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16483896 25 mg 25mg
16481498 5 mg 5mg
16472549 1 mg 1mg
16469088 10 mg 10mg
16434887 50 mg 50mg
5 options
This item is not returnable. View return policy

Product Code. 16483896

Brand: Medchem Express HYN0767

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Isoorientin is a potent inhibitor of COX-2 with an IC50 value of 39 μM.

Isoorientin is a potent inhibitor of COX-2 with an IC50 value of 39 μM.

Specifications

Chemical Name or Material Isoorientin
CAS 4261-42-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H20O11
Quantity 25 mg
Synonym Homoorientin
Solubility Information DMSO : ≥ 100 mg/mL (223.03 mM)
SMILES OC1=C(C2=O)C(OC(C3=CC(O)=C(O)C=C3)=C2)=CC(O)=C1[C@@H]([C@@H]([C@@H](O)[C@@H]4O)O)O[C@@H]4CO
Molecular Weight (g/mol) 448.38
Formula Weight 448.38
Percent Purity 99.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.