Learn More
Isonicotinoyl chloride hydrochloride, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 39178-35-3 |
| Molecular Formula | C6H5Cl2NO |
| MDL Number | MFCD00012830 |
| Quantity | 5 g |
| SMILES | O=C(Cl)C1=CC=NC=C1.[H]Cl |
| Molecular Weight (g/mol) | 178.02 |
| Percent Purity | 95% |
| Chemical Name or Material | Isonicotinoyl chloride hydrochloride |