Learn More
SymLine Isolating Amplifier
Product Code. 15922531
Change view
Click to view available options
For Use With (Equipment):
Signalbox T1, T5, Signalbox2
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15922531 Signalbox T1, T5, Signalbox2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15922531 Supplier SymLine Supplier No. 108278
  • ONLINE EXCLUSIVE PRICE
    4200.00 NOK
    3647.72 NOK / Pack of 1
    Save 552.28 NOK
    13% Off
Estimated Shipment: 18-12-2026
Switch amplifier for Signalbox devices.
  • Suitable for use with Signalbox T1, T5, Signalbox2
Specifications
Description Switch amplifier EU, 2 channels safety barrier, 120 - 230 V AC supply, contact or NAMUR-inputs / ATEX: Ex II (1) G [Ex ia] IIC, Ex II (1) D [Ex ia] IIIC
For Use With (Equipment) Signalbox T1, T5, Signalbox2
Product Type Switch Amplifier