Learn More
Isochlorogenic Acid A, TRC
Product Code. 30004288
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004288 5 mg 5mg
30006534 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30004288 Supplier Toronto Research Chemicals Supplier No. TRCI7897105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2450-53-5
Molecular Formula C25 H24 O12
Molecular Weight (g/mol) 516.45
Synonym Cyclohexanecarboxylic acid, 3,5-bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-1,4-dihydroxy-, (1α,3R,4α,5R)-, Cinnamic acid, 3,4-dihydroxy-, 5-carboxy-2,5-dihydroxy-1,3-cyclohexylene ester (8CI), Cyclohexanecarboxylic acid, 3,5-bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4-dihydroxy-, (1α,3R,4α,5R)- (9CI), Cyclohexanecarboxylic acid, 3,5-bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4-dihydroxy-, [1R-(1α,3α,4α,5β)]-, Cyclohexanecarboxylic acid, 1,3,4,5-tetrahydroxy-, 3,5-bis(3,4-dihydroxycinnamate) (8CI), (1α,3R,4α,5R)-3,5-Bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-1,4-dihydroxycyclohexanecarboxylic acid, 3,5-Dicaffeoylquinic acid, 3,5-Dicaffeylquinic acid, CJ 4-16-4, Isochlorogenic acid a, Quinic acid 3,5-di-O-caffeate
IUPAC Name (1R,3R,4S,5R)-3,5-bis[[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy]-1,4-dihydroxycyclohexane-1-carboxylic acid
SMILES O[C@@H]1[C@@H](C[C@](O)(C[C@H]1OC(=O)\C=C\c2ccc(O)c(O)c2)C(=O)O)OC(=O)\C=C\c3ccc(O)c(O)c3
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C25H24O12/c26-15-5-1-13(9-17(15)28)3-7-21(30)36-19-11-25(35,24(33)34)12-20(23(19)32)37-22(31)8-4-14-2-6-16(27)18(29)10-14/h1-10,19-20,23,26-29,32,35H,11-12H2,(H,33,34)/b7-3+,8-4+/t19-,20-,23-,25+/m1/s1
Formula Weight 516.1268
Percent Purity >95%
Chemical Name or Material Isochlorogenic Acid A
Grade HPLC