missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Isobutyltriphenylphosphonium bromide, 98+%, Thermo Scientific™

Product Code. 11443488
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11443488

Brand: Alfa Aesar™ A17881.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as intermediate for pharmaceutical.

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22884-29-3
Molecular Formula C22H24BrP
Molecular Weight (g/mol) 399.31
MDL Number MFCD00031593
InChI Key VIKPDPHNYHUZQW-UHFFFAOYSA-M
Synonym isobutyltriphenylphosphonium bromide, 2-methylpropyl triphenyl phosphanium bromide, phosphonium, 2-methylpropyl triphenyl-, bromide, 2-methylpropyl triphenylphosphanium bromide, acmc-1citv, isobutyltriphenylphosphoniumbromide, isobutyl triphenylphosphonium bromide, 2-methylpropyl triphenyl phosphonium bromide, isobutyl triphenyl phosphonium bromide, phosphonium, 2-methylpropyl triphenyl-,bromide 1:1
PubChem CID 2724571
SMILES [Br-].CC(C)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 198°C to 202°C
Quantity 5 g
Beilstein 4171228
IUPAC Name 2-methylpropyl(triphenyl)phosphanium;bromide
Formula Weight 399.32
Percent Purity ≥98%
Chemical Name or Material Isobutyltriphenylphosphonium bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.