Learn More
Isobutyl Methyl Ketone, Supelco™
Product Code. 15831201
Change view
Click to view available options
Quantity:
2.5 L
25 L
Unit Size:
2.5 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15831201 25 L 25 L
15826660 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15831201 Supplier Supelco Supplier No. 8208206025
 Restricted Item
Estimated Shipment: 19-11-2026
EMPLURA™
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108-10-1
Specifications
Melting Point −80°C (lit.)
Density 0.801 g/mL at 25°C
Quantity 25 L
Molecular Formula (CH3)2CHCH2COCH3
MDL Number MFCD00008938
Synonym 4-Methyl-2-pentanone, Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone, 4-Methyl-2-pentanone, Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone
Boiling Point 117°C to 118°C
Molecular Weight (g/mol) 100.16
Grade Research
Chemical Name or Material Isobutyl Methyl Ketone