Learn More
Isobutyl hexanoate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 105-79-3 |
| Molecular Formula | C10H20O2 |
| MDL Number | MFCD00048870 |
| Quantity | 25 g |
| SMILES | CCCCCC(OCC(C)C)=O |
| Molecular Weight (g/mol) | 172.26 |
| Percent Purity | 98% |
| Chemical Name or Material | Isobutyl hexanoate |