missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl Cinnamate, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 122-67-8 |
| Molecular Formula | C13H16O2 |
| Quantity | 100 g |
| Synonym | isobutyl cinnamate, labdanol, 2-methylpropyl cinnamate, cinnamic acid, isobutyl ester, 2-methylpropyl 3-phenylpropenoate, unii-4nci0mr7kp, isobutyl 3-phenylpropenoate, isobutyl beta-phenylacrylate, fema no. 2193, 2-methylpropyl 2e-3-phenylprop-2-enoate |
| InChI Key | IQZUZPKOFSOVET-CMDGGOBGSA-N |
| SMILES | CC(C)COC(=O)C=CC1=CC=CC=C1 |
| IUPAC Name | 2-methylpropyl (E)-3-phenylprop-2-enoate |
| Molecular Weight (g/mol) | 204.269 |
| PubChem CID | 778574 |
| Formula Weight | 204.25 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction