missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl chloroformate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Isobutyl chloroformate is used as a peptide reagent. It is also used in the preparation of phenethyl-carbamic acid isobutyl ester by reaction with phenethylamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isobutyl chloroformate is used as a peptide reagent. It is also used in the preparation of phenethyl-carbamic acid isobutyl ester by reaction with phenethylamine.
Solubility
Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, acids, bases, alcohols and amines. Moisture sensitive.
Isobutyl chloroformate is used as a peptide reagent. It is also used in the preparation of phenethyl-carbamic acid isobutyl ester by reaction with phenethylamine.
Solubility
Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, acids, bases, alcohols and amines. Moisture sensitive.
Specifications
Melting Point | -80°C |
Density | 1.048 |
Boiling Point | 128°C to 129°C |
Flash Point | 30°C (86°F) |
Assay Percent Range | 98% |
Linear Formula | ClCO2CH2CH(CH3)2 |
Refractive Index | 1.406 |
Quantity | 100 g |
UN Number | UN2742 |
Beilstein | 956590 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'