Learn More
Isobutyl Acetate, Sigma-Aldrich™
Product Code. 15871031
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15871031 100 mL 100mL
15826650 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15871031 Supplier Sigma Aldrich Supplier No. 8205570100
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-19-0
Specifications
Melting Point -99°C
Density 0.87 g/cm3 at 20°C
Boiling Point 116°C to 118°C/1013 hPa
Molecular Formula CH3COOCH2CH(CH3)2
MDL Number MFCD00008932
Quantity 100 mL
Synonym Isobutyl acetate, Acetic acid isobutyl ester, Isobutyl acetate
Molecular Weight (g/mol) 116.16
Grade Research