Learn More
Isobutanol, Supelco™
Product Code. 15834000
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15834000 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15834000 Supplier Supelco Supplier No. 1009842500
 Restricted Item
Estimated Shipment: 27-11-2026
For analysis EMSURE™ ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Specifications
Melting Point -108°C
CAS 78-83-1
Density 0.802 g/cm3 at 20°C
Quantity 2.5 L
Molecular Formula (CH3)2CHCH2OH
MDL Number MFCD00004740
Synonym Isobutanol, Isobutylalcohol, 2-Methyl-1-propanol, Iso-Butanol, Isobutanol
Boiling Point 106°C to 108°C/1013 hPa
Molecular Weight (g/mol) 74.12
Grade Research
Chemical Name or Material Isobutanol