Learn More
Isobornyl acetate, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | Isobornyl acetate |
| CAS | 125-12-2 |
| Density | 0.98 g/cm³ at 20°C |
| Boiling Point | 215°C (1013 hPa) |
| Flash Point | 89°C (closed cup) |
| Quantity | 250 mL |
| Synonym | Isobornyl acetate, Acetic acid isobornyl ester, exo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]hept-2-yl acetate, Isobornyl acetate |
| Molecular Weight (g/mol) | 196.29 |
| Grade | Not Available |