Learn More
Isoamyl Alcohol, Supelco™
Description
EMPLURA™ is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.
Specifications
Specifications
| CAS | 123-51-3 |
| Melting Point | -117°C |
| Density | 0.81 g/cm3 at 20°C |
| Boiling Point | 131°C/1013 hPa |
| MDL Number | MFCD00002977 |
| Quantity | 1 L |
| Synonym | Isoamyl alcohol, iso-Pentyl alcohol, 3-Methyl-1-butanol, Iso-Amylalcohol, Isoamyl alcohol |
| Molecular Weight (g/mol) | 88.15 |
| Grade | Research |