Learn More
Brady™ ISO Square Polypropylene Safety Sign 'Fire Alarm Flashing Light'
Product Code. 17851785
Change view
Click to view available options
Dimensions (L x W):
100 x 100 mm
148 x 148 mm
150 x 150 mm
200 x 200 mm
250 x 250 mm
315 x 315 mm
400 x 400 mm
Color:
Photoluminescent on Red
White on Red
Unit Size:
Each
Certifications/Compliance:
Class B
Class C
Product Code. Dimensions (L x W) Color unitSize Certifications/Compliance
17851785 400 x 400 mm White on Red Each -
17891735 100 x 100 mm White on Red Each -
17801745 148 x 148 mm White on Red Each -
17881735 200 x 200 mm White on Red Each -
17871735 250 x 250 mm White on Red Each -
17831785 315 x 315 mm White on Red Each -
17841745 100 x 100 mm Photoluminescent on Red Each Class B
17841705 100 x 100 mm Photoluminescent on Red Each Class C
17811695 150 x 150 mm Photoluminescent on Red Each Class B
17831745 150 x 150 mm Photoluminescent on Red Each Class C
17871705 200 x 200 mm Photoluminescent on Red Each Class B
17841785 200 x 200 mm Photoluminescent on Red Each Class C
12 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17851785
Brand: Brady™ 311210
Denna artikel kan inte returneras. Se returpolicy
ISO Safety Sign displaying flashing fire alarm and flames.
  • To indicate a device that flashes to indicate that the fire alarm has operated
Specifikationer
Alternate Part Number F/F018/NT/PP-400X400-1
Material Polypropylene
Quantity 1 Sign
Mounting Type Surface Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 15.75 in.
Width (English) 15.75 in.
Temperature Range (Metric) -40°C to +120°C
Shape Square
Temperature Range (English) -40°F to +248°F
Languages Universal
Dimensions (L x W) 400 x 400 mm
Displayed Text Fire Alarm Flashing Light
Product Type Safety Sign
Color White on Red
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.