Learn More
Brady™ ISO Square Polypropylene Safety Sign 'Fire Alarm Flashing Light'
Product Code. 17871735
Click to view available options
Dimensions (L x W):
100 x 100 mm
148 x 148 mm
150 x 150 mm
200 x 200 mm
250 x 250 mm
315 x 315 mm
400 x 400 mm
Color:
Photoluminescent on Red
White on Red
Unit Size:
Each
Certifications/Compliance:
Class B
Class C
Product Code. 17871735
Brand: Brady™ 311208
Only null left
Add to basket
Add to basket
ISO Safety Sign displaying flashing fire alarm and flames.
  • To indicate a device that flashes to indicate that the fire alarm has operated
Specifications
Alternate Part Number F/F018/NT/PP-250X250-1
Material Polypropylene
Quantity 1 Sign
Mounting Type Surface Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 9.84 in.
Width (English) 9.84 in.
Temperature Range (Metric) -40°C to +120°C
Shape Square
Temperature Range (English) -40°F to +248°F
Languages Universal
Dimensions (L x W) 250 x 250 mm
Displayed Text Fire Alarm Flashing Light
Product Type Safety Sign
Color White on Red