Learn More
Brady™ ISO Square Polypropylene Safety Sign 'Fire Alarm Flashing Light'
Product Code. 17841785
Change view
Click to view available options
Dimensions (L x W):
100 x 100 mm
148 x 148 mm
150 x 150 mm
200 x 200 mm
250 x 250 mm
315 x 315 mm
400 x 400 mm
Color:
Photoluminescent on Red
White on Red
Certifications/Compliance:
Class B
Class C
Unit Size:
Each
Product Code. Dimensions (L x W) Color Certifications/Compliance unitSize
17841785 200 x 200 mm Photoluminescent on Red Class C Each
17891735 100 x 100 mm White on Red - Each
17801745 148 x 148 mm White on Red - Each
17881735 200 x 200 mm White on Red - Each
17871735 250 x 250 mm White on Red - Each
17831785 315 x 315 mm White on Red - Each
17851785 400 x 400 mm White on Red - Each
17841745 100 x 100 mm Photoluminescent on Red Class B Each
17841705 100 x 100 mm Photoluminescent on Red Class C Each
17811695 150 x 150 mm Photoluminescent on Red Class B Each
17831745 150 x 150 mm Photoluminescent on Red Class C Each
17871705 200 x 200 mm Photoluminescent on Red Class B Each
12 options
Product Code. 17841785
Brand: Brady™ 311225
Only null left
Add to basket
Add to basket
ISO Safety Sign displaying flashing fire alarm and flames.
  • To indicate a device that flashes to indicate that the fire alarm has operated
Specifications
Certifications/Compliance Class C
Alternate Part Number F/F018/NT/PP-PHOLUMC-200X200-1
Finish Gloss
Material Polypropylene
Quantity 1 Sign
Mounting Type Surface Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 7.87 in.
Width (English) 7.87 in.
Temperature Range (Metric) -40°C to +66°C
Shape Square
Temperature Range (English) -40°F to +150°F
Languages Universal
Dimensions (L x W) 200 x 200 mm
Displayed Text Fire Alarm Flashing Light
Product Type Safety Sign
Color Photoluminescent on Red