Learn More
Brady™ ISO Safety Universal Sign 'Wear Laboratory Coat'
Product Code. 17841695
Change view
Click to view available options
Dimensions (L x W):
10 x 10 cm
15 x 15 cm
20 x 20 cm
31.5 x 31.5 cm
5 x 5 cm
Material:
Polyester
Polypropylene
Unit Size:
Each
Pack of 2
Product Code. Dimensions (L x W) Material unitSize
17841695 10 x 10 cm Polypropylene Each
17861775 5 x 5 cm Polyester Pack of 2
17871775 10 x 10 cm Polyester Each
17881775 15 x 15 cm Polyester Each
17861745 15 x 15 cm Polypropylene Each
17821725 20 x 20 cm Polyester Each
17861785 20 x 20 cm Polypropylene Each
17891695 31.5 x 31.5 cm Polyester Each
17851745 31.5 x 31.5 cm Polypropylene Each
9 options
Product Code. 17841695
Brand: Brady™ 311369
Only null left
Add to basket
Add to basket
Safety Sign to signify that a laboratory coat must be worn.
  • Provide visual warning that laboratory coat must be worn in the area
Specifications
Alternate Part Number M/M059/NT/PP-DIA100-1
Material Polypropylene
Quantity 1 Sign
Size (English) 3.94 in. Dia.
Diameter (Metric) 100 mm
Mounting Type Surface Mount
For Use With (Application) Facility Identification, Safety Identification
Thickness 1.55 mm
Temperature Range (Metric) -40°C to +120°C
Shape Round
Temperature Range (English) -40°F to +248°F
Languages Universal
Dimensions (L x W) 10 x 10 cm
Displayed Text Wear Laboratory Coat
Product Type Safety Sign
Color White on Blue