Learn More
Scharlab ISO-OKTAN PA 1L
Brand:  Scharlab IS01541000
ISO-OKTAN PA 1L
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11368614
  • ONLINE EXCLUSIVE PRICE
    1022.70 NOK
    915.84 NOK / 1 L
    Save 106.86 NOK
    10% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 2,2,4-Trimetheylpentane, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .