Learn More
Brady™ ISO 7010 Sign - Fire hose reel
Product Code. 15614377
Change view
Click to view available options
Height (Metric):
148 mm
200 mm
315 mm
Width (Metric):
148 mm
200 mm
315 mm
Unit Size:
Each
Product Code. Height (Metric) Width (Metric) unitSize
15614377 315 mm 315 mm Each
15684367 148 mm 148 mm Each
15694367 200 mm 200 mm Each
3 options
Product Code. 15614377
Brand: Brady™ 817050
Only null left
Add to basket
Add to basket
Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC F002-315x315-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 315 mm
Thickness 1.501 mm
Width (Metric) 315 mm
Temperature Range (Metric) -40°C to 120°C
Shape Square
Languages Global
Color White on Red