Learn More
Brady™ ISO 7010 Sign - Fire extinguisher
Product Code. 15684357 Shop All Brady Products
Change view
Click to view available options
Height (Metric):
100 mm
148 mm
200 mm
315 mm
Width (Metric):
100 mm
148 mm
200 mm
315 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) Unit Size
15684357 315 mm 315 mm Each
15644357 100 mm 100 mm Each
15654357 148 mm 148 mm Each
15664357 200 mm 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15684357 Supplier Brady™ Supplier No. 816891
  • ONLINE EXCLUSIVE PRICE
    171.00 NOK
    162.90 NOK / Pack of 1
    Save 8.10 NOK
    4% Off
Estimated Shipment: 18-11-2026
Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC F001-315x315-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 315 mm
Thickness 1.501 mm
Width (Metric) 315 mm
Temperature Range (Metric) -40°C to 120°C
Shape Square
Languages Global
Color White on Red