Learn More
ISO 22032 PBDE Stock Standard Mixture 360 50 μg/mL in Isooctane:Toluene, Dr. Ehrenstorfer
Product Code. 30313056
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313056 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30313056 Supplier Dr. Ehrenstorfer Supplier No. DREA50000360IT
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 189084-64-8, 68631-49-2, 207122-15-4, 207122-16-5, 5436-43-1, 60348-60-9
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Isooctane/Toluene