missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Islet 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15661-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the LIM/homeodomain family of transcription factors. The encoded protein binds to the enhancer region of the insulin gene, among others, and may play an important role in regulating insulin gene expression. The encoded protein is central to the development of pancreatic cell lineages and may also be required for motor neuron generation. Mutations in this gene have been associated with maturity-onset diabetes of the young.Specifications
| Islet 1 | |
| Polyclonal | |
| Unconjugated | |
| ISL1 | |
| Isl 1, ISL LIM homeobox 1, ISL1, Islet 1, ISLET1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16392, 3670, 64444 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61371, P61372, P61374 | |
| Isl1 | |
| Islet 1 Fusion Protein Ag8088 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction