Learn More
Isepamicin sulfate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 67814-76-0 |
| Molecular Formula | C22H45N5O16S |
| MDL Number | MFCD01683537 |
| Quantity | 25 mg |
| SMILES | O=C(N[C@H]1[C@H](O[C@@H]2[C@H](O)[C@@H](NC)[C@@](C)(O)CO2)[C@@H](O)[C@H](O[C@@H]3[C@H](O)[C@@H](O)[C@H](O)[C@@H](CN)O3)[C@@H](N)C1)[C@@H](O)CN.O=S(O)(O)=O |
| Molecular Weight (g/mol) | 667.68 |
| Percent Purity | 98% |
| Chemical Name or Material | Isepamicin sulfate |