Learn More
Micronova™ Irradiated Vinyl Cleanroom Tape
BIOPHARMA
Product Code. 12833582 Shop All Micronova Products
Change view
Click to view available options
Color:
Light Blue
Transparent
White
Dimensions (L x W):
36 yd. x 0.5 in.
36 yd. x 0.75 in.
36 yd. x 1 in.
36 yd. x 2 in.
Unit Size:
16 rolls
Each
Pack of 16
Pack of 24
Pack of 36
Pack of 48
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) unitSize
12833582 Light Blue 36 yd. x 1 in. Pack of 36
15388690 Transparent 36 yd. x 0.75 in. 16 rolls
17808359 Transparent 36 yd. x 0.75 in. Pack of 16
17103915 White 36 yd. x 0.5 in. Each
12743376 White 36 yd. x 0.75 in. Pack of 48
12853542 White 36 yd. x 1 in. Pack of 36
12883542 White 36 yd. x 2 in. Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12833582 Supplier Micronova™ Supplier No. CR1001LBIR
  • ONLINE EXCLUSIVE PRICE
    10160.00 NOK
    9838.70 NOK / Pack of 36
    Save 321.30 NOK
    3% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
Irradiated vinyl cleanroom tape, with rubber, clean-peel adhesive, is extremely conformable and ideal for media preparation, batching, color-coding and packaging in critical and aseptic processing applications.
Irradiated vinyl cleanroom tape is designed to limit particulate and bio-burden counts in critical and aseptic processing applications. This extremely conformable tape is ideal for sealing curved and irregular surfaces and the synthetic rubber adhesive ensures clean peel from wafer cartridges and most cleanroom work surfaces.
  • Irradiated, ultra processed, pressure-sensitive vinyl tape
  • Low particulate and bio-burden counts
  • Synthetic rubber adhesive, ensures clean peel from most surfaces
  • Extremely conformable; tapes and seals curved and irregular surfaces
  • Ideal for batching, color coding, packaging, etc. in aseptic areas
  • Excellent for clean construction areas
  • Can be written on with cleanroom pen
  • Cleanroom processed and wound on a plastic core
  • Supplied with full certification of irradiation and lot traceability
  • Individually double bagged
  • Available in Transparent, White and Light Blue
  • Available in 1/2, 3/4, 1 and 2-inch of widths
  • Roll length: 33 m (36 yd.)
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 Certified Facility, Certificate of Irradiation, Certificate of Conformance
Adhesive Type Rubber
Coating Vinyl
Color Light Blue
Finish Matte
Format Tape
Material Vinyl
Product Type Cleanroom Tape
Sterility Sterile
For Use With (Application) Plate sealing, marking and color coding in aseptic environments.
Quantity 36/Cs.
Temperature Range (English) 59°F to 77°F
Temperature Range (Metric) 15°C to 25°C
Dimensions (L x W) 36 yd. x 1 in.
Length (English) 36 yd.
Length (Metric) 33 m
Width (English) 1 in.
Width (Metric) 25.4 mm
Width (Metric) Tape 25.4 mm