Description
The Fisherbrand Irradiated Polyethylene Cleanroom Tape is an excellent general purpose tape for media preparation, batching, color coding, packaging, and other applications within the lab and sterile processing areas.
- Irradiated, pressure-sensitive polyethylene tape
- Ideal for sealing, color coding and marking in aseptic areas
- Excellent for clean construction areas
- Cleanroom processed and wound on a 3-inch plastic core
- Supplied with full certification of irradiation and lot traceability
- Individually double bagged
- Available in a range of widths
- Roll length: 33 m (36 yd.)
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Adhesive Type | Acrylic |
| Color | White |
| Material | Polyethylene |
| Sterility | Sterile |
| For Use With (Application) | Plate sealing, marking and color coding in aseptic environments |
| Temperature Range (English) | 60°F to 80°F |
| Temperature Range (Metric) | 16°C to 27°C |
| Dimensions (L x W) | 33 m x 25.4 mm (36 yd. x 1 in.) |
| Length (English) | 36 yd. |
| Show More |