Learn More
Fisherbrand™ Irradiated Cleanroom Vinyl Tape
Product Code. 16285162 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Transparent
White
Dimensions (LxW):
36 yd. x 0.5 in.
36 yd. x 0.5 in.)
36 yd. x 0.75 in.
36 yd. x 1 in.
36 yd. x 2 in.
Unit Size:
Pack of 16
Pack of 24
Pack of 36
Product Code. Color Dimensions (LxW) unitSize
16285162 Transparent 36 yd. x 0.75 in. Pack of 16
16275162 White 36 yd. x 0.5 in.) Pack of 24
16235162 White 36 yd. x 0.5 in. Pack of 24
16255162 White 36 yd. x 1 in. Pack of 36
16295162 White 36 yd. x 0.75 in. Pack of 16
16225162 Transparent 36 yd. x 0.5 in. Pack of 24
16245162 Transparent 36 yd. x 1 in. Pack of 36
16265162 Transparent 36 yd. x 2 in. Pack of 24
8 options
Product Code. 16285162
Irradiated vinyl cleanroom tape with low particulate and bio-burden counts that is ideal for media preparation, batching, color coding, packaging, and other applications within the lab, sterile processing and clean construction areas.
  • Irradiated vinyl cleanroom tape forms a strong bond with plastics, polymers and stainless steel without leaving adhesive residue behind.
  • Processed in a clean environment, this tape is designed for light industrial use, clean construction, and sterile processing areas.
  • Conformable with rubber adhesive.
  • Forms a strong bond with plastics, polymers, and stainless steel.
  • Does not leave adhesive residue when removed.
  • Ideal for media preparation, batching, color coding with cleanroom pen.
  • 3 inch plastic core.
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Processing, NIKA - FDA, CID, Complies with Federal Specification PPP-T-66Eand CID A-A 1689A
Adhesive Type Rubber
Coating Vinyl
Color Transparent
Finish Semi-gloss
Format Roll
Material Vinyl
Product Type Cleanroom Tape
Sterility Sterile
For Use With (Application) Plate sealing, marking and color coding in aseptic environments
Includes 3 in. Plastic Core
Quantity 16/Cs.
Size (English) 8 x 8 x 8 in.
Size (Metric) 20.32 x 20.32 x 20.32 cm
Temperature Range (English) 60°F to 80°F
Temperature Range (Metric) 16°C to 27°C
Dimensions (LxW) 36 yd. x 0.75 in.
Length (English) 36 yd.
Length (Metric) 33 m
Width (English) 0.75 in.
Width (Metric) 19.05 mm