Learn More
Fisherbrand™ Irradiated Cleanroom Vinyl Tape
Product Code. 16275162 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Transparent
White
Dimensions (L x W):
36 yd. x 0.5 in.
36 yd. x 0.5 in.)
36 yd. x 0.75 in.
36 yd. x 1 in.
36 yd. x 2 in.
Unit Size:
Pack of 16
Pack of 24
Pack of 36
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) unitSize
16275162 White 36 yd. x 0.5 in.) Pack of 24
16235162 White 36 yd. x 0.5 in. Pack of 24
16255162 White 36 yd. x 1 in. Pack of 36
16295162 White 36 yd. x 0.75 in. Pack of 16
16225162 Transparent 36 yd. x 0.5 in. Pack of 24
16285162 Transparent 36 yd. x 0.75 in. Pack of 16
16245162 Transparent 36 yd. x 1 in. Pack of 36
16265162 Transparent 36 yd. x 2 in. Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 16275162
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
Irradiated vinyl cleanroom tape with low particulate and bio-burden counts that is ideal for media preparation, batching, color coding, packaging, and other applications within the lab, sterile processing and clean construction areas.
  • Irradiated vinyl cleanroom tape forms a strong bond with plastics, polymers and stainless steel without leaving adhesive residue behind.
  • Processed in a clean environment, this tape is designed for light industrial use, clean construction, and sterile processing areas.
  • Conformable with rubber adhesive.
  • Forms a strong bond with plastics, polymers, and stainless steel.
  • Does not leave adhesive residue when removed.
  • Ideal for media preparation, batching, color coding with cleanroom pen.
  • 3 inch plastic core.
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Processing, NIKA - FDA, CID, Complies with Federal Specification PPP-T-66Eand CID A-A 1689A
Description Cleanroom 1/2 inch White Vinyl Tape
Adhesive Type Rubber
Coating Vinyl
Color White
Finish Matte White
Format Roll
Material Vinyl
Product Type Cleanroom Tape
Sterility Sterile
For Use With (Application) Sealing, Labeling, Color Coding, Floor Marking
Includes 3 in. Plastic Core
Quantity 72/Cs.
Size Case
Size (English) 16 x 10 x 6 in.
Size (Metric) 40.6 x 25.4 x 15.24 cm
Temperature Range (English) 60°F to 80°F
Temperature Range (Metric) 16°C to 27°C
Dimensions (L x W) 36 yd. x 0.5 in.)
Length (English) 36 yd.
Length (Metric) 33 m
Width (English) 0.5 in.
Width (Metric) 1.3 cm