Learn More
Iron(II) fumarate, 94%
Product Code. 11436836
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
Product Code. Quantity unitSize
11436836 250 g 250g
11426836 1000 g 1000g
2 options
Product Code. 11436836
Brand: Thermo Scientific Alfa Aesar B23956.30
Request Bulk or Custom Format
CAS: 141-01-5 | C4H2FeO4-2 | 169.901 g/mol
Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.
Solubility
Soluble in water (0.14 g/100 ml at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 141-01-5
Molecular Formula C4H2FeO4-2
Molecular Weight (g/mol) 169.901
MDL Number MFCD00058315
InChI Key OOPLWEDSEDELIX-TYYBGVCCSA-L
Synonym ferrous fumarate, eisenfumarat
PubChem CID 66524104
IUPAC Name (E)-but-2-enedioate;iron
SMILES C(=CC(=O)[O-])C(=O)[O-].[Fe]
Specifications
Melting Point >300°C
Quantity 250 g
Merck Index 14,4046
Solubility Information Soluble in water (0.14g/100 ml at 25°C).
Formula Weight 169.9
Percent Purity 94%
Density 2.44
Chemical Name or Material Iron(II) fumarate