missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) fluoride, anhydrous, 98%

Product Code. p-7048790
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11377147

Brand: Thermo Scientific Alfa Aesar 011486.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-28-8 | F2Fe | 93.84 g/mol

Iron(II) fluoride is used for proteomics research applications. It is also a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-28-8
Molecular Formula F2Fe
Molecular Weight (g/mol) 93.84
MDL Number MFCD00016093
InChI Key FZGIHSNZYGFUGM-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) difluoride
SMILES [F-].[F-].[Fe++]

Specifications

Melting Point >1000°C (sublimation)
Color White
Physical Form Powder
Quantity 10 g
UN Number UN3260
Merck Index 14,4045
Solubility Information Insoluble in water.
Formula Weight 93.84
Percent Purity 98%
Sensitivity Air sensitive; Hygroscopic
Density 4.09 g/mL
Chemical Name or Material Iron(II) fluoride, Anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Iron(II) fluoride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.