Learn More
Iron(II) fluoride, anhydrous, 98%
Product Code. 11387147
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11387147
Brand: Thermo Scientific Alfa Aesar 011486.18
Request Bulk or Custom Format
CAS: 7789-28-8 | F2Fe | 93.84 g/mol
Iron(II) fluoride is used for proteomics research applications. It is also a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7789-28-8
Molecular Formula F2Fe
Molecular Weight (g/mol) 93.84
MDL Number MFCD00016093
InChI Key FZGIHSNZYGFUGM-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) difluoride
SMILES [F-].[F-].[Fe++]
Specifications
Melting Point >1000°C (sublimation)
Color White
Physical Form Powder
Quantity 50 g
UN Number UN3260
Merck Index 14,4045
Solubility Information Insoluble in water.
Formula Weight 93.84
Percent Purity 98%
Sensitivity Air sensitive; Hygroscopic
Density 4.09 g/mL
Chemical Name or Material Iron(II) fluoride, Anhydrous