Learn More
Iron(II) acetate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 3094-87-9 |
| Molecular Formula | C4H6FeO4 |
| MDL Number | MFCD00058909 |
| Quantity | 500 g |
| SMILES | CC([O-])=O.CC([O-])=O.[Fe+2] |
| Molecular Weight (g/mol) | 173.93 |
| Percent Purity | 95% |
| Chemical Name or Material | Iron(II) acetate |